missing translation for 'onlineSavingsMsg'
Learn More

Basic Fuchsin Sulfate (Technical Grade), TRC

Product Code. 30137196
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137196 5 g 5g
30135514 10 g 10g
17934435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137196 Supplier Toronto Research Chemicals Supplier No. TRCB1187235G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20 H19 N3 . XH2SO4
Molecular Weight (g/mol) 399.5
Synonym C.I. Basic Violet 14, Monohydrochloride, Rosaniline, 12418 Red, Aizen Magenta, Astra Fuchsine B, Basic Fuchsine, Basic Magenta, Basic Magenta E 200, Basic Violet 14, C-WR Violet 8, C.I. 42510, C.I. Basic Violet 14, Calcozine Fuchsine HO, Calcozine Magenta RTN, Calcozine Magenta XX, Cerise B, Diabasic Magenta, Diamond Fuchsine, Fuchsin, Fuchsin Basic, Fuchsine, Fuchsine A, Fuchsine CS, Fuchsine G, Fuchsine HO, Fuchsine N, Fuchsine RTN, Fuchsine SBP, Fuchsine Y, Lowacryl Violet 14, Magenta DP, Magenta E, Magenta G, Magenta I, Magenta PN, Magenta Powder N, Magenta S, Magenta Superfine, Orient Basic Magenta, RGB 20, RGN 10, RGN 10 (dye), Rosaniline Chloride, Rosaniline Hydrochloride, Rosanilinium Chloride, 4-[(4-aminophenyl)(4-imino-2,5-cyclohexadien-1-ylidene)methyl]-2-methyl-Benzenamine Monohydrochloride
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-2-methyl-aniline;sulfuric acid
SMILES Cc1cc(ccc1N)C(=C2C=CC(=N)C=C2)c3ccc(N)cc3.OS(=O)(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C20H19N3.H2O4S/c1-13-12-16(6-11-19(13)23)20(14-2-7-17(21)8-3-14)15-4-9-18(22)10-5-15;1-5(2,3)4/h2-12,21H,22-23H2,1H3;(H2,1,2,3,4)/b20-14-,21-17?;
Chemical Name or Material Basic Fuchsin Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.