Learn More
Buerkle™ Basket for Sample Bottles Vampire
Shop All Bürkle Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
Diameter 100 mm.
Specifications
Specifications
| Product Type | Bottle Holder |
| Material | Stainless Steel |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Sterility | Non-sterile |
| For Use With (Application) | For use with Buerkle Vampire Sampler |
| For Use With (Equipment) | Bottle holder |