missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BASP1 Antibody (10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26902CL1
This item is not returnable.
View return policy
Description
BASP1 Monoclonal antibody specifically detects BASP1 in Human samples. It is validated for ELISA
Specifications
| BASP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| brain abundant, membrane attached signal protein 1,22 kDa neuronal tissue-enriched acidic protein, brain acid-soluble protein 1, CAP-23, CAP23brain acid soluble protein 1, NAP-22, NAP22MGC8555, Neuronal axonal membrane protein NAP-22, neuronal tissue-enriched acidic protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BASP1 (Uniprot#: P80723; aa3-227). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10409 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction