missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BASP1 Recombinant Rabbit Monoclonal Antibody (5H3L1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703692
This item is not returnable.
View return policy
Description
This antibody is predicted to react with Mouse, Rat, Bovine.
BASP1 (brain acid soluble protein 1) is a membrane bound protein with several transient phosphorylation sites and PEST motifs. Conservation of proteins with PEST sequences among different species supports their functional significance. PEST sequences typically occur in proteins with high turnover rates. Immunological characteristics of this protein are species specific. This protein also undergoes N-terminal myristoylation.
Specifications
| BASP1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P80723 | |
| BASP1 | |
| Peptides corresponding to human BASP1 [1) aa7-aa24 2) aa138-aa155]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 5H3L1 | |
| Unconjugated | |
| BASP1 | |
| 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10409 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction