missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bassoon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114895
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Bassoon.
Bassoon is an approximately 420 kDa protein that is a localized at the presynaptic nerve terminals and is believed to play a role in the structural and functional organization of the synaptic vesicle cycle. Bassoon does not belong to any known protein families. It has been found in rat and mouse with sequence identity of the two proteins at 96%. The human BASSOON gene has recently been cloned and localized. Bassoon is predicted to contain two double-zinc fingers, three coiled-coil regions, and two polyglutamine domains. The polyglutamine domains in the C-terminus are of interest, since it is known that for some human proteins, such as Huntingtin, abnormal amplification of this region can cause late-onset neurodegeneration. Bassoon is concentrated at sites opposite to postsynaptic densities in synaptic terminals and in cultured neurons, it is found to colocalize with GABA (A) and glutamate (GluR1) receptors. Another presynaptic protein, Piccolo was found to colocalize with Bassoon in cultured hippocampal neurons. These observations suggested that they serve specific functions at synaptic junctions and may be involved in organization of the cytoskeleton at the site of neurotransmitter release.
Specifications
| Bassoon | |
| Polyclonal | |
| Unconjugated | |
| BSN | |
| bassoon; bassoon (presynaptic cytomatrix protein); bassoon presynaptic cytomatrix protein; BSN; Kiaa0434; neuronal double zinc finger protein; presynaptic cytomatrix protein; protein bassoon; Zinc finger protein 231; ZNF231 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12217, 29138, 8927 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O88737, O88778, Q9UPA5 | |
| BSN | |
| A synthesized peptide derived from human BSN(Accession Q9UPA5), corresponding to amino acid residues R3594-E3644. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction