Learn More
Bassoon Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 30392-1-AP-20UL
 View more versions of this product
Product Code. 18088105
  • € 145.00 / 20µL
Description
The Bassoon antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Bassoon. This antibody recognizes human, mouse antigen. The Bassoon antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Bassoon
550 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunohistochemistry, ELISA
BSN, KIAA0434, Protein bassoon, Zinc finger protein 231, ZNF231
20 μL
8927
Store at -20°C. Stable for one year after shipment.