missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bat Immunoglobulin Antibody (BT1-4F10), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23483APCCY7
This item is not returnable.
View return policy
Description
Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for Flow Cytometry.
Specifications
| Bat Immunoglobulin | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Microchiropteran bat (Eptesicus fuscus) spleen cells. | |
| 0.1 mL | |
| Immunoglobulin L (Light) chain. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| BT1-4F10 | |
| Flow Cytometry | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Bat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction