missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41191H
This item is not returnable.
View return policy
Description
BATF Polyclonal antibody specifically detects BATF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| BATF | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BATF. The immunogen is located within the first 50 amino acids of BATF. Amino Acid Squence: SFSRSPPPGKQDSSDD | |
| 0.1 mL | |
| Signal Transduction | |
| 10538 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction