missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BATF3 Antibody (841702) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7437A
This item is not returnable.
View return policy
Description
BATF3 Monoclonal antibody specifically detects BATF3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| BATF3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 841702 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT21-kD small nuclear factor isolated from T cells | |
| E. coli-derived recombinant human BATF3, Met1-Arg127, Accession # Q9NR55 | |
| 0.1 mL | |
| Immunology | |
| 55509 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction