Learn More
Julabo™ Bath Adapters
Click to view available options
Size:
G1 x 1/4 in. F to BF 1/2 in. ID
M10 x 1 to M16 x 1
M16 x 1 F to NPT 1/2 in. F
M16 x 1 female to 2 x M16 x 1 male
M16 x 1 female to M16 x 1 female
M16 x 1 female to NPT 1/2 in. male
M16 x 1 female to NPT 1/4 in. male
M16 x 1 female to NPT 3/8 in. male
M16 x 1 female to tube 1/4 in. male
M16 x 1 female to tube 3/8 in. male
M16 x 1 male to NPT 1/4 in. female
M16 x 1 male/male
M24 x 1/2 F to NPT 1/2 in. F
M24 x 1/2 F to NPT 3/8 in. M
M24 x 1/2 in. F to M16 x 1 M
M24 x 1/2 male to M24 x 1/2 male
Unit Size:
Each
Pack of 2
Specifications
Specifications
| For Use With (Equipment) | CF30, CF31, CF40, CF41 Cryo-compact circulator, F25-HE refrigerated/heating circulator |
| Type | Adapter |
| Size | M16 x 1 male/male |