missing translation for 'onlineSavingsMsg'
Learn More

Bavachinin, MedChemExpress

Product Code. 16458908
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16458908

Brand: Medchem Express HYN0234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bavachinin(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae;has potent anti-angiogenic activity.

Bavachinin(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae

Specifications

Chemical Name or Material Bavachinin
CAS 19879-30-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C21H22O4
Quantity 10 mg
Synonym 7-O-Methylbavachin Bavachinin A
Solubility Information DMSO : 125 mg/mL (369.39 mM; Need ultrasonic)
SMILES O=C1C[C@@H](C2=CC=C(O)C=C2)OC3=CC(OC)=C(C/C=C(C)\C)C=C13
Molecular Weight (g/mol) 338.4
Formula Weight 338.4
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.