missing translation for 'onlineSavingsMsg'
Learn More

Bavisant dihydrochloride hydrate, MedChemExpress

Product Code. 16402367
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16402367

Brand: Medchem Express HY14880B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bavisant dihydrochloride hydrate (JNJ31001074AAC) is an orally active, potent, brain-penetrating and highly selective antagonist of the histamine H3 receptor. Bavisant dihydrochloride hydrate can be used for attention-deficit hyperactivity disorder (ADHD) research.

Bavisant dihydrochloride hydrate (JNJ31001074AAC) is an orally active, potent, brain-penetrating and highly selective antagonist of the histamine H3 receptor. Bavisant dihydrochloride hydrate can be used for attention-deficit hyperactivity disorder (ADHD) research.

Specifications

Chemical Name or Material Bavisant dihydrochloride hydrate
Physical Form Solid
CAS 1103522-80-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H31Cl2N3O3
For Use With (Application) Neuroscience-Neuromodulation
Synonym JNJ31001074AACJNJ31001074 dihydrochloride hydrate
Solubility Information H2O : ≥ 50 mg/mL (118.94 mM) ∣DMSO : ≥ 1 mg/mL (2.38 mM)
SMILES O=C(N1CCN(C2CC2)CC1)C(C=C3)=CC=C3CN4CCOCC4.[H]Cl.[H]Cl.O
Molecular Weight (g/mol) 420.37
Formula Weight 420.37
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.