missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax Antibody (CL7919) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21162MFV500
This item is not returnable.
View return policy
Description
Bax Monoclonal antibody specifically detects Bax in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Bax | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
| This antibody was developed using a synthetic peptide derived from Q07812, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 581 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7919 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction