Learn More
Bax inhibitor 1 Antibody (20F565), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-24912AF700
Product Code. 18186872
  • € 559.00 / 0.10mL
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bax inhibitor 1
Monoclonal
Alexa Fluor 700
P55061
TMBIM6
A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061.
0.1 mL
Apoptosis
7009
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry (Paraffin)
20F565
Western Blot, Immunohistochemistry-Paraffin
BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6
Mouse
Protein G purified
RUO
Primary
This antibody is reactive to Human, Mouse, Rat.
Store at 4C in the dark.
IgG2b κ