missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 091

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BAY 091 is a potent and selective, ATP-dependent, phosphatidylinositol-5-phosphate 4-kinase type-2 alpha (PIP4K2A) inhibitor (IC50 values are 1.3 nM, 8.5 nM and 16.4 nM with 10 μM ATP, or in an HTRF assay with 10 μM and 2 mM ATP, respectively).
Specifications
Specifications
| Chemical Name or Material | (R)-2-((3-Cyano-2-(2 '-fluoro-3 '-methyl-[1,1 '-biphenyl]-4-yl)-1,7-naphthyridin-4-yl)amino)butanoic acid |
| CAS | 2922280-34-8 |
| Quantity | 50 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C26H21FN4O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction