Learn More
BAY-1436032, MedChemExpress
Product Code. 16485487
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16485487
Brand: Medchem Express HY100020
MedChemExpress BAY-1436032 is a novel pan-mutant isocitrate dehydrogenase 1 (IDH1) inhibitor.
BAY-1436032 is a novel pan-mutant isocitrate dehydrogenase 1 (IDH1) inhibitor.
Specifications
Chemical Name or Material BAY-1436032
CAS 1803274-65-8
Color Gray
Purity Grade Notes Research
Molecular Formula C26H30F3N3O3
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (255.35 mM; Need ultrasonic and warming)
SMILES O=C(O)CCC1=CC=C2C(N=C(NC3=CC=C(OC(F)(F)F)C=C3)N2[C@H]4CC(C)(C)C[C@@H](C)C4)=C1
Molecular Weight (g/mol) 489.53
Formula Weight 489.53
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only