missing translation for 'onlineSavingsMsg'
Learn More

BAY-299, MedChemExpress

Product Code. 16476827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476827

Brand: Medchem Express HY107424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY-299 is a very potent, dual inhibitor with IC50s of 67 nM for BRPF2 bromodomains (BD), 8 nM for TAF1 BD2, and 106 nM for TAF1L BD2.

BAY-299 is a very potent, dual inhibitor with IC50s of 67 nM for BRPF2 bromodomains (BD), 8 nM for TAF1 BD2, and 106 nM for TAF1L BD2.

Specifications

Chemical Name or Material BAY-299
Physical Form Solid
CAS 2080306-23-4
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H23N3O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 25 mg/mL (58.21 mM; Need ultrasonic and warming)
SMILES O=C(N1C2=C(C)C=C(N(C)C3=O)C(N3C)=C2)C4=CC=C(CCCO)C5=C4C(C1=O)=CC=C5
Molecular Weight (g/mol) 429.47
Formula Weight 429.47
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.