missing translation for 'onlineSavingsMsg'
Learn More

BAY 57-1293 Methanesulfonate Salt, TRC

Product Code. 30240503
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240503 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30240503 Supplier Toronto Research Chemicals Supplier No. TRCB1188055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1428333-96-3
Molecular Formula C18H18N4O3S2 . CH4O3S
Quantity 5 mg
Synonym BAY 57-1293 Methanesulfonate Salt, N-[5-(Aminosulfonyl)-4-methyl-1,3-thiazol-2-yl]-N-methyl-2-[4-(2-pyridinyl)phenyl]acetamide Methanesulfonate Salt, Pritelivir Methanesulfonate;
InChI Formula InChI=1S/C18H18N4O3S2.CH4O3S/c1-12-17(27(19,24)25)26-18(21-12)22(2)16(23)11-13-6-8-14(9-7-13)15-5-3-4-10-20-15;1-5(2,3)4/h3-10H,11H2,1-2H3,(H2,19,24,25);1H3,(H,2,3,4)
SMILES CN(C(=O)Cc1ccc(cc1)c2ccccn2)c3nc(C)c(s3)S(=O)(=O)N.CS(=O)(=O)O
IUPAC Name methanesulfonic acid;N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-(4-pyridin-2-ylphenyl)acetamide
Molecular Weight (g/mol) 498.6
Chemical Name or Material N-[5-(Aminosulfonyl)-4-methyl-2-thiazolyl]-N-methyl-4-(2-pyridinyl)-benzeneacetamide Methanesulfonate Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.