Learn More
Bay 59-3074, MedChemExpress
Product Code. 16492586
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16492586
Brand: Medchem Express HY100488
MedChemExpress Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist with Ki values of 48.3 and 45.5 nM at human CB1 and CB2 receptors, respectively. Bay 59-3074 has analgesic properties.
Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist with Ki values of 48.3 and 45.5 nM at human CB1 and CB2 receptors, respectively. Bay 59-3074 has analgesic properties.
Specifications
Chemical Name or Material Bay 59-3074
CAS 406205-74-1
Color White
Purity Grade Notes Research
Molecular Formula C18H13F6NO4S
Quantity 25 mg
Solubility Information DMSO : ≥ 34 mg/mL (75.00 mM)
SMILES O=S(CCCC(F)(F)F)(OC1=CC=CC(OC2=CC=CC(C(F)(F)F)=C2C#N)=C1)=O
Molecular Weight (g/mol) 453.36
Formula Weight 453.36
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only