missing translation for 'onlineSavingsMsg'
Learn More

Bay 59-3074, MedChemExpress

Product Code. p-200097050
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16492586

Brand: Medchem Express HY100488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist with Ki values of 48.3 and 45.5 nM at human CB1 and CB2 receptors, respectively. Bay 59-3074 has analgesic properties.

Bay 59-3074 is a selective cannabinoid CB1/CB2 receptor partial agonist with Ki values of 48.3 and 45.5 nM at human CB1 and CB2 receptors, respectively. Bay 59-3074 has analgesic properties.

Specifications

Chemical Name or Material Bay 59-3074
CAS 406205-74-1
Color White
Purity Grade Notes Research
Molecular Formula C18H13F6NO4S
Quantity 25 mg
Solubility Information DMSO : ≥ 34 mg/mL (75.00 mM)
SMILES O=S(CCCC(F)(F)F)(OC1=CC=CC(OC2=CC=CC(C(F)(F)F)=C2C#N)=C1)=O
Molecular Weight (g/mol) 453.36
Formula Weight 453.36
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.