missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 805

Product Code. p-200112510
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30306028

Brand: Tocris Bioscience™ 7984/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective inhibitor of USP21; also NF-κB activator

BAY 805 is a potent and selective inhibitor of USP21 (IC50 = 6 nM, Kd = 2.2 nM). BAY 805 induces NF-κB activation (EC50= 17 nM in a cell-based reporter assay).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(1R)-1-[[[5-[(4-Cyanophenyl)methyl]-1,3,4-thiadiazol-2-yl]amino]carbonyl]-3-methylbutyl]-1-(trifluoromethyl)cyclohexanecarboxamide
CAS 2925481-88-3
Quantity 10 mg
Target Deubiquitinating Enzyme Inhibitors
Molecular Formula C24H28F3N5O2S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.