missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBOX1 Antibody (OTI4F9), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71372DL594
This item is not returnable.
View return policy
Description
BBOX1 Monoclonal antibody specifically detects BBOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
Specifications
| BBOX1 | |
| Monoclonal | |
| DyLight 594 | |
| 2-oxoglutarate dioxygenase, 2-oxoglutarate dioxygenase 1, BBHgamma-butyrobetaine, BBOXEC 1.14.11.1, butyrobetaine (gamma), 2-oxoglutarate dioxygenase (gamma-butyrobetainehydroxylase) 1, Gamma-BBH, Gamma-butyrobetaine, gamma-butyrobetaine dioxygenase, Gamma-butyrobetaine hydroxylase, G-BBH | |
| Full length recombinant protein of human BBOX1 (NP_003977) produced in HEK293T cells. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
| 8424 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunofluorescence, Immunofluorescence | |
| OTI4F9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction