Learn More
Brady™ BBP12 Label Printer Kit EU with Unwinder and Workstation LAB Suite
This kit contains the BBP12 Label Printer, unwinder, and the Workstation Laboratory Identification Software suite so that you can immediately start designing professional labels.
Brand: Brady™ 306866
Additional Details : Weight : 5.30000kg
Description
The Brady™ BBP™12 gives you the versatility to print all your basic label types from one machine, saving costs, reducing downtime and simplifying your labeling.
- Over 800 varieties of labels, including external-feed bulk rolls and custom parts
- Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
- Centering ribbon spindle virtually eliminates ribbon wrinkle
- Compact size won't clutter your workplace or bench
- Comes with unwinder and Brady™ Workstation LAB Suite
Specifications
Color LCD | |
8 in. | |
7.5 in. | |
10 in. | |
Not Autoclavable | |
Monochrome | |
Czech, Dutch, Norwegian, Polish, French, German, Greek, Hungarian, Swedish, Kazakh, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh | |
BBP12 | |
300 dpi | |
Windows™ 7, Windows™ 8, Windows™ 10 | |
5.1 lb. | |
Healthcare and Hospital Labeling, Laboratory Labeling | |
110/240 V |
Thermal Transfer | |
203 mm | |
191 mm | |
259 mm | |
Non-sterile | |
BBP12 Printer, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Brady™ Workstation Laboratory ID Suite | |
128 MB (Flash), 64 MB (SDRAM), SD Flash Memory Card Up To 32 GB | |
101.6 mm/s | |
Ethernet, Serial, USB | |
5°C to 40°C | |
2.3 kg | |
1 Year |