missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BBS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545025
This item is not returnable.
View return policy
Description
Peptide sequence: GSDLFWTVTG DNVNSLALCD FDGDGKKELL VGSEDFDIRV FKEDEIVAEM Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
This gene is a member of the Bardet-Biedl syndrome (BBS) gene family. Bardet-Biedl syndrome is an autosomal recessive disorder characterized by severe pigmentary retinopathy, obesity, polydactyly, renal malformation and mental retardation. The proteins encoded by BBS gene family members are structurally diverse and the similar phenotypes exhibited by mutations in BBS gene family members is likely due to their shared roles in cilia formation and function. Many BBS proteins localize to the basal bodies, ciliary axonemes, and pericentriolar regions of cells. BBS proteins may also be involved in intracellular trafficking via microtubule-related transport. The protein encoded by this gene forms a multiprotein BBSome complex with six other BBS proteins.
Specifications
| BBS2 | |
| Polyclonal | |
| Unconjugated | |
| BBS2 | |
| 2410125H22Rik; AI447581; Bardet-Biedl syndrome 2; Bardet-Biedl syndrome 2 (human); Bardet-Biedl syndrome 2 homolog; Bardet-Biedl syndrome 2 protein; Bardet-Biedl syndrome 2 protein homolog; BBS; bbs2; bbs2 protein; fb80a05; MGC20703; RP74; Unknown (protein for MGC:134372); wu:fb80a05 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 583 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BXC9 | |
| BBS2 | |
| Synthetic peptide directed towards the N-terminal of human BBS2 (aa 151-200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction