missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS4 Antibody (OTI2D5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70241AF532
This item is not returnable.
View return policy
Description
BBS4 Monoclonal specifically detects BBS4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BBS4 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BBS4 | |
| Recombinant protein fragment corresponding to amino acids 266-519 of human BBS4 (NP_149017) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI2D5 | |
| Western Blot | |
| Bardet-Biedl syndrome 4, Bardet-Biedl syndrome 4 protein | |
| Mouse | |
| 0.1 mL | |
| 585 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction