missing translation for 'onlineSavingsMsg'
Learn More

BBS5 Antibody (7E11) [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30518517 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30518517 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30518517 Supplier Novus Biologicals Supplier No. NBP342934JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BBS5 Monoclonal antibody specifically detects BBS5 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BBS5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 700000000000
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Bardet-Biedl syndrome 5, Bardet-Biedl syndrome 5 protein, DKFZp762I194
Host Species Mouse
Immunogen Bardet-Biedl syndrome 5 - (Uniprot# Q8N3I7)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 129880
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.