Learn More
Invitrogen™ BBS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113979
Description
Immunogen sequence: VEIDSDGHTD AFVAYFADGN KQQDREPVFS EELGLAIEKL KDGFTLQGLW For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.
The BBSome complex is thought to function as a coat complex required for sorting of specific membrane proteins to the primary cilia. The BBSome complex is required for ciliogenesis but is dispensable for centriolar satellite function. This ciliogenic function is mediated in part by the Rab8 GDP/GTP exchange factor, which localizes to the basal body and contacts the BBSome. Rab8(GTP) enters the primary cilium and promotes extension of the ciliary membrane. Firstly the BBSome associates with the ciliary membrane and binds to RAB3IP/Rabin8, the guanosyl exchange factor (GEF) for Rab8 and then the Rab8-GTP localizes to the cilium and promotes docking and fusion of carrier vesicles to the base of the ciliary membrane. The BBSome complex, together with the LTZL1, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. Required for BBSome complex ciliary localization but not for the proper complex assembly.
Specifications
| BBS5 | |
| Polyclonal | |
| Unconjugated | |
| BBS5 | |
| 1700049I01Rik; 2700023J09Rik; Bardet-Biedl syndrome 5; Bardet-Biedl syndrome 5 (human); Bardet-Biedl syndrome 5 homolog; Bardet-Biedl syndrome 5 protein; Bardet-Biedl syndrome 5 protein homolog; Bbs5; novel DUF1448 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 129880 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N3I7 | |
| BBS5 | |
| Synthetic peptide directed towards the middle region of human BBS5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.