missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBX Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71906AF700
This item is not returnable.
View return policy
Description
BBX Polyclonal antibody specifically detects BBX in Human samples. It is validated for Western Blot
Specifications
| BBX | |
| Polyclonal | |
| Western Blot | |
| Bobby sox homolog, bobby sox homolog (Drosophila), HBP2HSPC339, HMG box transcription factor BBX, HMG box-containing protein 2, MDS001, x 001 protein | |
| The immunogen for this product maps to a region between residue 891 and 941 of human Bobby Sox Homolog using the numbering given in entry NP_001136040.1 (GeneID 56987). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 56987 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction