missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCA3 Antibody (OTI1E3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72057AF405
This item is not returnable.
View return policy
Description
BCA3 Monoclonal specifically detects BCA3 in Human samples. It is validated for Western Blot.
Specifications
| BCA3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AKIP1 | |
| Full length recombinant protein of human BCA3 (NP_878904) produced in E.coli. | |
| 0.1 mL | |
| 56672.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1E3 | |
| Western Blot | |
| AKIP1koyt binding protein 2, BCA3koyt binding protein 3, breast cancer associated gene 3, Breast cancer-associated gene 3 protein, chromosome 11 open reading frame 17, koyt binding protein 1, PKA-interacting protein, proline-rich protein BCA3, protein kinase A-interacting protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction