missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAM/CD239 Antibody (002), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89356APC
This item is not returnable.
View return policy
Description
BCAM/CD239 Monoclonal antibody specifically detects BCAM/CD239 in Human samples. It is validated for ELISA, ELISA
Specifications
| BCAM/CD239 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 002 | |
| ELISA, Sandwich ELISA Detection | |
| antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM/CD239 (Accession#: NP_005572.2; Met1-Ala547). | |
| 0.1 mL | |
| Cardiovascular Biology, CD Markers, Immunology | |
| 4059 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction