missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcan Mouse anti-Human, Rat, FITC, Clone:S294A-6, Abnova™
Mouse monoclonal antibody raised against partial recombinant rat Bcan.
Brand: Abnova MAB16839.100ug
This item is not returnable.
View return policy
Specifications
| Bcan | |
| Monoclonal | |
| FITC | |
| Immunocytochemistry Immunofluorescence Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Bcan | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| S294A-6 | |
| brevican | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| ALPBRE | |
| Recombinant protein corresponding to amino acids 219-655 of rat Bcan. | |
| 25393 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction