Learn More
BCAP29 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand:  Abnova H00055973-AP11.1Set
Product Code. 16153534
  • € 843.00 / 1 set
Specifications
BCAP29
Unconjugated
BAP29/DKFZp686M2086
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-BCAP29 (100 ug) 2. Detection antibody: mouse monoclonal anti-BCAP29, IgG2a Kappa (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
B-cell receptor-associated protein 29
BCAP29
RUO
55973
Human
Ab Pair