missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-681MFV500
This item is not returnable.
View return policy
Description
BCAS1 Polyclonal antibody specifically detects BCAS1 in Human samples. It is validated for Western Blot, Western Blot
Specifications
| BCAS1 | |
| Polyclonal | |
| Western Blot, Simple Western | |
| breast carcinoma amplified sequence 1, breast carcinoma-amplified sequence 1, NABC1AIBC1Amplified and overexpressed in breast cancer, Novel amplified in breast cancer 1 | |
| Reacts with residues 532-550 [DTNSLQNGDKLQKRPEKRQ] of human NABC1. | |
| 0.1 mL | |
| Cancer, Prostate Cancer | |
| 8537 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction