missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAS2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77124JF646
This item is not returnable.
View return policy
Description
BCAS2 Polyclonal antibody specifically detects BCAS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| BCAS2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| breast carcinoma amplified sequence 2, Breast carcinoma-amplified sequence 2, DAM1DNA amplified in mammary carcinoma 1 protein, pre-mRNA-splicing factor SPF27, Snt309, SPF27, spliceosome associated protein, amplified in breast cancer, Spliceosome-associated protein SPF 27 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human BCAS2. The immunogen is located within the last 50 amino acids of BCAS2. Amino Acid Squence: QHGEANKENIRQDF | |
| 0.1 mL | |
| Cancer | |
| 10286 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction