missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCAS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520905
This item is not returnable.
View return policy
Description
A suggested positive control is MCF7 cell lysate. PA5-20905 can be used with blocking peptide PEP-1019.
cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1.5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.
Specifications
| BCAS2 | |
| Polyclonal | |
| Unconjugated | |
| BCAS2 | |
| 6430539P16Rik; AI132645; BCAS2; breast carcinoma amplified sequence 2; breast carcinoma-amplified sequence 2; Breast carcinoma-amplified sequence 2 homolog; C76366; C80030; DAM1; DNA amplified in mammary carcinoma 1 protein; pre-mRNA-splicing factor SPF27; Snt309; SPF27; spliceosome associated protein, amplified in breast cancer; spliceosome-associated protein SPF 27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10286, 295334, 68183 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O75934, Q9D287 | |
| BCAS2 | |
| A 14 amino acid peptide near the carboxy terminus of human BCAS2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction