missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCAS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584771
This item is not returnable.
View return policy
Description
Immunogen sequence: MAGTGLVAGE VVVDALPYFD QGYEAPGVRE AAAALVEEET RRYRPTKNYL S Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1.5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.
Specifications
| BCAS2 | |
| Polyclonal | |
| Unconjugated | |
| BCAS2 | |
| 6430539P16Rik; AI132645; BCAS2; breast carcinoma amplified sequence 2; breast carcinoma-amplified sequence 2; Breast carcinoma-amplified sequence 2 homolog; C76366; C80030; DAM1; DNA amplified in mammary carcinoma 1 protein; pre-mRNA-splicing factor SPF27; Snt309; SPF27; spliceosome associated protein, amplified in breast cancer; spliceosome-associated protein SPF 27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75934 | |
| BCAS2 | |
| Recombinant protein corresponding to Human BCAS2. Recombinant protein control fragment (Product #RP-104167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction