missing translation for 'onlineSavingsMsg'
Learn More

BCAS2 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Product Code. p-7087964 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16806793 20 μL 20µL
16896783 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16806793 Supplier Proteintech Supplier No. 104141AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

cDNA encoding BCAS2 has been isolated. The deduced 225-amino acid protein contains a central potential N-glycosylation site and multiple N-terminal potential phosphorylation sites. Northern and Southern blot analyses demonstrates expression of a 1. 5-kb transcript at various levels in multiple breast cancer cell lines, with greatest amplification in BT-20 and MCF-7 cells. BCAS2 is ubiquitously expressed in normal tissues.

Specifications

Antigen BCAS2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BCAS2
Gene Accession No. O75934, Q9D287
Gene Alias BCAS2, DAM1, Pre mRNA splicing factor SPF27
Gene Symbols BCAS2
Host Species Rabbit
Immunogen BCAS2 Fusion Protein Ag0669
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10286, 295334, 68183
Target Species Mouse, Human, Rat, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.