missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCAT1 Antibody (OTI3F5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70242AF700
This item is not returnable.
View return policy
Description
BCAT1 Monoclonal specifically detects BCAT1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BCAT1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BCAT1 | |
| Full length recombinant protein of human BCAT1 (NP_005495) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI3F5 | |
| Western Blot, Immunohistochemistry | |
| BCAT(c), BCATC, BCT1PNAS121, branched chain amino-acid transaminase 1, cytosolic, branched chain aminotransferase 1, cytosolic, branched-chain-amino-acid aminotransferase, cytosolic, DKFZp686E12175, EC 2.6.1.42, ECA39, MECA39, placental protein 18, PP18, Protein ECA39 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 586.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction