missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress BCDA

Product Code. 16491936
Encompass_Preferred
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 16491936

Brand: MedChemExpress HY52112

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BCDA (5-bromo-4-chloroindoxyl acetate) is a chromogenic substrate of esterase used to potently detect the activity of esterase.

Specifications

CAS 3252-36-6
Solubility Ethanol : ≥ 100 mg/mL (346.58 mM)
Synonym 5-bromo-4-chloroindoxyl acetate
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Quantity 500 mg
Molecular Weight (g/mol) 288.53 g/mol
Product Type Dye Reagents
Purity or Quality Grade 99.0%
Shipping Condition Room temperature
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.