Learn More
MedChemExpress BCIP
Specifications
Specifications
| CAS | 6578-06-9 |
| Solubility | DMSO : ≥ 51 mg/mL (117.61 mM) ∣DMF : 25 mg/mL (57.65 mM∣Need ultrasonic) |
| Synonym | BCIP p-toluidine salt∣ X-phosphate p-toluidine salt |
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 433.62 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 98.0% |
| Shipping Condition | Room temperature |