missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDHB Polyclonal Antibody

BCKDHB Polyclonal Antibody
Brand: Invitrogen PA587963
This item is not returnable.
View return policy
Description
BCKDHB Polyclonal Antibody for Western Blot
Specifications
| BCKDHB | |
| Polyclonal | |
| Unconjugated | |
| Bckdhb | |
| 2-oxoisovalerate dehydrogenase beta subunit; 2-oxoisovalerate dehydrogenase subunit beta, mitochondrial; BCKD BCKDH E1-beta; BCKDE1B; BCKDH E1-beta; BCKDHB; branched chain alpha-ketoacid dehydrogenase E1-beta subunit; branched chain keto acid dehydrogenase E1 beta; branched chain keto acid dehydrogenase E1 subunit beta; branched chain keto acid dehydrogenase E1, beta polypeptide; branched chain ketoacid dehydrogenase E1, beta polypeptide; branched-chain alpha-keto acid dehydrogenase E1 component beta chain; dJ279A18.1; E1B; E1b-beta subunit of the branched-chain complex; testis secretory sperm-binding protein Li 240mP | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12040, 29711 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35738, Q6P3A8 | |
| Bckdhb | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 183-392 of human BCKDHB (NP_8988711). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction