missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCKDK Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99564CL1
This item is not returnable.
View return policy
Description
BCKDK Polyclonal antibody specifically detects BCKDK in Human samples. It is validated for Western Blot
Specifications
| BCKDK | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| BCKDHKIN, branched chain alpha-ketoacid dehydrogenase kinase, branched chain ketoacid dehydrogenase kinase, EC 2.7.11, EC 2.7.11.4, mitochondrial | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human BCKDK. | |
| 0.1 mL | |
| Protein Kinase | |
| 10295 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction