missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-215AF405
This item is not returnable.
View return policy
Description
Bcl-10 Polyclonal antibody specifically detects Bcl-10 in Human samples. It is validated for Western Blot
Specifications
| Bcl-10 | |
| Polyclonal | |
| B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB | |
| 8915 | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| Immunogen affinity purified | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction