missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcl-10 Monoclonal Antibody (3C11F4)
GREENER_CHOICE

Product Code. 17460773 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17460773 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17460773 Supplier Invitrogen™ Supplier No. MA538455

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-10
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3C11F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BCL10
Gene Accession No. O95999
Gene Alias AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1
Gene Symbols BCL10
Host Species Mouse
Immunogen Purified recombinant fragment of human BCL10 (aa: 98-234) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.