missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Antibody (BCL2/782 + BCL2/796), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47853DL594
This item is not returnable.
View return policy
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Bcl-2 | |
| Monoclonal | |
| DyLight 594 | |
| apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
| Recombinant full-length human bcl-2 protein (Uniprot: P10415) | |
| 0.1 mL | |
| Apoptosis, Autophagy, BiologicAll speciesy Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Oncogenes, Tumor Suppressors | |
| 596 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| BCL2/782 + BCL2/796 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction