missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Mouse anti-Human, Clone: rBCL6/1718, Novus Biologicals™
Mouse Monoclonal Antibody
€ 499.00
Specifications
| Antigen | Bcl-6 |
|---|---|
| Clone | rBCL6/1718 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
Bcl-6 Monoclonal specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Bcl-6 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 604 | |
| IgG1 κ | |
| Protein A or G purified |
| rBCL6/1718 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
| Recombinant human Bcl-6 protein fragment (aa256-389) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title