missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl G Antibody (OTI6D1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72204H
This item is not returnable.
View return policy
Description
Bcl G Monoclonal specifically detects Bcl G in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Bcl G | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BCL2L14 | |
| Recombinant protein fragment corresponding to amino acids 116-216 of human Bcl G (NP_620048) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI6D1 | |
| Western Blot, Immunohistochemistry | |
| apoptosis facilitator Bcl-2-like protein 14, Apoptosis regulator Bcl-G, Bcl2-L-14, BCL2-like 14 (apoptosis facilitator), BCL-G, BCLGbcl2-L-14 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 79370.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction