missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-G Monoclonal Antibody (OTI6D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527070
This item is not returnable.
View return policy
Description
Bcl-G Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
| Bcl-G | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZR8 | |
| BCL2L14 | |
| Human recombinant protein fragment corresponding to amino acids 116-216 of BCL2L14 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6D1 | |
| Unconjugated | |
| BCL2L14 | |
| 4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79370 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction