missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Antibody (BCL2L1/2406), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08777AF350
This item is not returnable.
View return policy
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| bcl-x | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Apoptosis regulator Bcl-X;BCL2L;BCL2L1;Bcl-2-like protein 1;BCLX;Bcl-X;BCL-XL/S;PPP1R52 | |
| Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary) (Uniprot: Q07817 ) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| BCL2L1/2406 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 598 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction