missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL Antibody (016), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89507UV
This item is not returnable.
View return policy
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Bcl-xL | |
| Monoclonal | |
| DyLight 350 | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Bcl-xL (NP_612815.1; Met1-Arg212). | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
| 598 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 16 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction